Magnesium chloride hexahydrate
Supplier: Roth Carl
Synonyms: Magnesium dichloride
3532.1 3532.2 3532.3
ROTH3532.1EA 0 EUR
ROTH3532.1 ROTH3532.2 ROTH3532.3
Magnesium chloride hexahydrate
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684