Magnesium chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Magnesium dichloride
M-Clarity™ quality level = MQ200
M2393-100G M2393-500G
SIALM2393-100GEA 53.3 EUR
SIALM2393-100G SIALM2393-500G
Magnesium chloride hexahydrate, Sigma-Aldrich®
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684