Magnesium sulphate heptahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
63138-1KG 63138-250G
SIAL63138-1KGEA 258 EUR
SIAL63138-1KG SIAL63138-250G
Magnesium sulphate heptahydrate, Sigma-Aldrich®
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715