Magnesium sulphate heptahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
M-Clarity™ quality level = MQ200
M2773-1KG M2773-500G
SIALM2773-1KGEA 253 EUR
SIALM2773-1KG SIALM2773-500G
Magnesium sulphate heptahydrate, Sigma-Aldrich®
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
Density: 2.66 g/cm³ (20 °C)
MDL Number: MFCD00149785
CAS Number: 10034-99-8
Merck Index: 13,05715