Aluminium nitrate nonahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Aluminum nitrate nonahydrate
M-Clarity™ quality level = MQ200
229415-100G 229415-10G
SIAL229415-100GEA 548 EUR
SIAL229415-100G SIAL229415-10G
Aluminium nitrate nonahydrate, Sigma-Aldrich®
Aluminium nitrate nonahydrate
Formula: Al(NO₃)₃·9H₂O
MW: 375.13 g/mol
Melting Pt: 73
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS Number: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1 III