Ammonium heptamolybdate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate
M-Clarity™ quality level = MQ200
09878-100G 09878-25G 09878-500G
SIAL09878-100GEA 112 EUR
SIAL09878-100G SIAL09878-25G SIAL09878-500G
Ammonium heptamolybdate tetrahydrate, Sigma-Aldrich®
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536