Ammonium iron(III) sulphate dodecahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Alum iron, Ammonium iron sulphate dodecahydrate, Ferric ammonium sulphate dodecahydrate, Ammonium iron(III) sulphate dodecahydrate, Iron alum, Ammonium ferric sulphate dodecahydrate
F3629-250G F3629-500G
SIALF3629-250GEA 44.6 EUR
SIALF3629-250G SIALF3629-500G
Ammonium iron(III) sulphate dodecahydrate, Sigma-Aldrich®
Ammonium iron (III) sulphate dodecahydrate
Formula: NH₄Fe(SO₄)₂·12H₂O
MW: 482.2 g/mol
Boiling Pt: 230
Melting Pt: 39…41 °C
Density: 1.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549