Gadolinium trichloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Gadolinium trichloride hexahydrate
G7532-5G G7532-25G
SIALG7532-5GEA 84.1 EUR
SIALG7532-5G SIALG7532-25G
Gadolinium trichloride hexahydrate, Sigma-Aldrich®
Gadolinium trichloride hexahydrate
Formula: Cl₃Gd·6H₂O
MW: 371.7 g/mol
MDL Number: MFCD00149732
CAS Number: 13450-84-5
EINECS: 233-386-6