Lanthanum(III) chloride heptahydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
203521-100G 203521-25G
SIAL203521-100GEA 353 EUR
SIAL203521-100G SIAL203521-25G
Lanthanum(III) chloride heptahydrate, Sigma-Aldrich®
Lanthanum (III) chloride heptahydrate
Formula: LaCl₃·7H₂O
Boiling Pt: ∼860 °C (1013 hPa)
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5