Cerium trinitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate
M-Clarity™ quality level = MQ100
392219-100G 392219-25G
SIAL392219-100GEA 486 EUR
SIAL392219-100G SIAL392219-25G
Cerium trinitrate hexahydrate, Sigma-Aldrich®
Cerium trinitrate hexahydrate
Formula: Ce(NO₃)₃·6H₂O
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 39 °C
Density: 2.377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1,II