ESD safety shoes, sandals, Velcro® fastening, uvex 1, 8536
Supplier: UVEX
8536.2/35 8536.2/36 8536.2/37 8536.2/38 8536.2/39 8536.2/40 8536.2/41 8536.2/42 8536.2/43 8536.2/44 8536.2/45 8536.2/46 8536.2/47 8536.2/48 8536.2/49 8536.2/50 8536.2/51 8536.2/52 8536.3/43 8536.3/42
114-6781EA 146 EUR
114-6781 114-6782 114-6783 114-6784 114-6785 114-6786 114-6787 114-6788 114-6789 114-6790 114-6791 114-6792 114-6793 114-6794 114-6795 114-6796 114-6797 114-6798 UVEX8536.3/43 UVEX8536.3/42
ESD safety shoes, sandals, Velcro® fastening, uvex 1, 8536
Shoes and Boots
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable water resistant high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Flexible, metal-free puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/red
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1P, SRC