Copper(II) chloride dihydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate
MQ200
C3279-100G C3279-500G
SIALC3279-100GEA 69.2 EUR
SIALC3279-100G SIALC3279-500G
Copper(II) chloride dihydrate, Sigma-Aldrich®
Copper (II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8 III
Merck Index: 13,02660