Copper(II) nitrate trihydrate
Supplier: Roth Carl
Danger
Synonyms: Cupric nitrate trihydrate, Copper dinitrate trihydrate
0255.1 0255.2 0255.3
ROTH0255.1EA 0 EUR
ROTH0255.1 ROTH0255.2 ROTH0255.3
Copper(II) nitrate trihydrate
Copper (II) nitrate trihydrate
Formula: Cu(NO₃)₂·3H₂O
Melting Pt: 114 °C
Density: 2.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149671
CAS Number: 10031-43-3
EINECS: 221-838-5
UN: 1477
ADR: 5.1,II
Merck Index: 13,02671