Copper(II) sulphate pentahydrate
Supplier: Roth Carl
Warning
Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate
P024.1 P024.2
ROTHP024.1EA 36 EUR
ROTHP024.1 ROTHP024.2
Copper(II) sulphate pentahydrate
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 13,02682