Copper(II) sulphate pentahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate
M-Clarity™ quality level = MQ200
469130-250G 469130-50G
SIAL469130-250GEA 365 EUR
SIAL469130-250G SIAL469130-50G
Copper(II) sulphate pentahydrate, Sigma-Aldrich®
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 13,02682