Yttrium(III) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Yttrium trichloride hexahydrate
M-Clarity™ quality level = MQ100
211648-250G 211648-50G
SIAL211648-250GEA 794 EUR
SIAL211648-250G SIAL211648-50G
Yttrium(III) chloride hexahydrate, Sigma-Aldrich®
Yttrium (III) chloride hexahydrate
Formula: YCl₃·6H₂O
MW: 303.36 g/mol
Boiling Pt: 1507
Melting Pt: 153
Density: 2.18 g/cm³ (20 °C)
MDL Number: MFCD00149941
CAS Number: 10025-94-2
EINECS: 233-801-0