Yttrium(III) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Yttrium trichloride hexahydrate
M-Clarity™ quality level = MQ100
464317-100G 464317-25G
SIAL464317-100GEA 292 EUR
SIAL464317-100G SIAL464317-25G
Yttrium(III) chloride hexahydrate, Sigma-Aldrich®
Yttrium (III) chloride hexahydrate
Formula: YCl₃·6H₂O
MW: 303.36 g/mol
Boiling Pt: 1507
Melting Pt: 153
Density: 2.18 g/cm³ (20 °C)
MDL Number: MFCD00149941
CAS Number: 10025-94-2
EINECS: 233-801-0