Cobalt(II) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate
M-Clarity™ quality level = MQ300
C8661-100G C8661-25G
SIALC8661-100GEA 191 EUR
SIALC8661-100G SIALC8661-25G
Cobalt(II) chloride hexahydrate, Sigma-Aldrich®
Cobalt (II) chloride hexahydrate
Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: 110
Melting Pt: 56
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 13,02462