Nickel(II) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nickel dichloride hexahydrate
M-Clarity™ quality level = MQ100
203866-5G 203866-25G
SIAL203866-5GEA 54.9 EUR
SIAL203866-5G SIAL203866-25G
Nickel(II) chloride hexahydrate, Sigma-Aldrich®
Nickel (II) chloride hexahydrate
Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
Melting Pt: 140
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 13,06529