Cobalt(II) nitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate
M-Clarity™ quality level = MQ100
203106-10G 203106-50G
SIAL203106-10GEA 195 EUR
SIAL203106-10G SIAL203106-50G
Cobalt(II) nitrate hexahydrate, Sigma-Aldrich®
Cobalt (II) nitrate hexahydrate
Formula: Co(NO₃)₂·6H₂O
Melting Pt: 57 °C
Density: 1.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,III
Merck Index: 13,02469