Iron(III) nitrate nonahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate
M-Clarity™ quality level = MQ100
254223-10G 254223-250G 254223-50G
SIAL254223-10GEA 98.4 EUR
SIAL254223-10G SIAL254223-250G SIAL254223-50G
Iron(III) nitrate nonahydrate, Sigma-Aldrich®
Iron (III) nitrate nonahydrate
Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125
Melting Pt: 47
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1 III
Merck Index: 13,04057