Iron(II) sulphate heptahydrate
Supplier: Roth Carl
Warning
Synonyms: Iron vitriol, Ferrous sulphate heptahydrate
P015.1 P015.2 P015.5
ROTHP015.1EA 42.2 EUR
ROTHP015.1 ROTHP015.2 ROTHP015.5
Iron(II) sulphate heptahydrate
Iron (II) sulphate heptahydrate
Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091