Nickel sulphate heptahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nickel (II) sulphate heptahydrate
M-Clarity™ quality level = MQ200
203890-10G 203890-50G
SIAL203890-10GEA 74.8 EUR
SIAL203890-10G SIAL203890-50G
Nickel sulphate heptahydrate, Sigma-Aldrich®
Nickel sulphate heptahydrate
Formula: NiO₄S·7H₂O
MW: 280.86 g/mol
Melting Pt: 99
Density: ∼2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149814
CAS Number: 10101-98-1
EINECS: 232-104-9
UN: 3077
ADR: 9 III