N-Acetyl-L-cysteine, Sigma-Aldrich®

Supplier: Merck

Synonyms: Acetylcysteine, N-Acetyl-3-mercaptoalanine, NAC, (R)-(+)-N-Acetylcysteine, (R)-2-Acetamido-3-mercaptopropanoic acid, Ac-Cys-OH, N-Acetyl-L-Cysteine, LNAC

M-Clarity™ quality level = MQ200

A8199-100G A8199-10G
SIALA8199-100GEA 612 EUR
SIALA8199-100G SIALA8199-10G
N-Acetyl-L-cysteine, Sigma-Aldrich®
N-Acetyl-L-cysteine
Formula: C₅H₉NO₃S
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 106…108 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004880
CAS Number: 616-91-1
EINECS: 210-498-3
Merck Index: 13,00090

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR