L(+)-Cysteine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
30089-100G
30089-25G
30089-500G
SIAL30089-100GEA
251
EUR
SIAL30089-100G
SIAL30089-25G
SIAL30089-500G
L(+)-Cysteine, Sigma-Aldrich®
L-Cysteine
Formula:
HSCH₂CH(NH₂)CO₂H Boiling Pt: 254 °C (1013 hPa) Melting Pt: 115 °C Density: 1.523 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064306 CAS Number: 52-90-4 EINECS: 200-158-2 Merck Index: 13,02810 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...