L(+)-Cysteine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ300
W326305-100G
W326305-1KG
W326305-5KG
W326305-SAMPLE
SIALW326305-100GEA
76.6
EUR
SIALW326305-100G
SIALW326305-1KG
SIALW326305-5KG
SIALW326305-SAMPLE
L(+)-Cysteine, Sigma-Aldrich®
L-Cysteine
Formula:
HSCH₂CH(NH₂)CO₂H Boiling Pt: 254 °C (1013 hPa) Melting Pt: 115 °C Density: 1.523 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064306 CAS Number: 52-90-4 EINECS: 200-158-2 Merck Index: 13,02810 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...