Potassium-D-gluconate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
M-Clarity™ quality level = MQ200
P1847-1KG P1847-500G
SIALP1847-1KGEA 385 EUR
SIALP1847-1KG SIALP1847-500G
Potassium-D-gluconate, Sigma-Aldrich®
Potassium D-gluconate
Formula: HOH₂C(CH(OH))₄COOK
MW: 234.25 g/mol
Melting Pt: 174…176 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00064211
CAS Number: 299-27-4
EINECS: 206-074-2
Merck Index: 12,07796