L(-)-Malic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
M7397-100G M7397-25G M7397-500G
SIALM7397-100GEA 239 EUR
SIALM7397-100G SIALM7397-25G SIALM7397-500G
L(-)-Malic acid, Sigma-Aldrich®
L(-)-Malic acid
Formula: C₄H₆O₅
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 101…104 °C
Density: 1.595 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064213
CAS Number: 97-67-6
EINECS: 202-601-5
Merck Index: 13,05730