(2R,3R)-(-)-2,3-Butanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: D(-)-2,3-Butanediol, (2R,3R)-Butane-2,3-diol, (R,R)-(-)-2,3-Butylene glykol, (R,R)-(-)-2,3-Butanediol, (2R,3R)-(-)-2,3-Dihydroxy butane, (R,R)-(-)-2,3-Dihydroxy butane, (2R,3R)-(-)-2,3-Butylene glykol
M-Clarity™ quality level = MQ200
237639-1G 237639-5G
SIAL237639-1GEA 74.3 EUR
SIAL237639-1G SIAL237639-5G
(2R,3R)-(-)-2,3-Butanediol, Sigma-Aldrich®
(2R,3R)-(-)-2,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 175
Melting Pt: 16
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064267
CAS Number: 24347-58-8
EINECS: 246-186-9
Merck Index: 13,01567