D-(-)-Sorbitol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
M-Clarity™ quality level = MQ200
85529-1KG 85529-250G 85529-5KG
SIAL85529-1KGEA 352 EUR
SIAL85529-1KG SIAL85529-250G SIAL85529-5KG
D-(-)-Sorbitol, Sigma-Aldrich®
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 94…98 °C
Density: 1.524 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM