D-(-)-Leucine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
H-D-Leu-OH, (R)-(-)-Leucine
M-Clarity™ quality level = MQ100
855448-10G
855448-2.5G
855448-25G
SIAL855448-10GEA
120
EUR
SIAL855448-10G
SIAL855448-2.5G
SIAL855448-25G
D-(-)-Leucine, Sigma-Aldrich®
D(-)-Leucine
Formula:
C₆H₁₃NO₂ Boiling Pt: 226 °C (1013 hPa) |
MDL Number:
MFCD00063088 CAS Number: 328-38-1 EINECS: 206-327-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...