(R)-(-)-2-Amino-3-methyl-1-butanol, Sigma-Aldrich®

Supplier: Merck

Synonyms: D(-)-2-Amino-3-methyl-1-butanol, (R)-(-)-2-Amino-3-methylbutanol, D-Valinol, D(-)-Valinol

M-Clarity™ quality level = MQ100

284483-1G 284483-5G
SIAL284483-1GEA 25.6 EUR
SIAL284483-1G SIAL284483-5G
(R)-(-)-2-Amino-3-methyl-1-butanol, Sigma-Aldrich®
(R)-(-)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Density: 0.93 g/cm³ (20 °C)
MDL Number: MFCD00064297
CAS Number: 4276-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR