L(+)-Histidine, Sigma-Aldrich®
Supplier: Merck
Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
M-Clarity™ quality level = MQ200
H8000-100G H8000-10G H8000-1KG H8000-25G H8000-500G H8000-5G H8000-5KG
SIALH8000-100GEA 177 EUR
SIALH8000-100G SIALH8000-10G SIALH8000-1KG SIALH8000-25G SIALH8000-500G SIALH8000-5G SIALH8000-5KG
L(+)-Histidine, Sigma-Aldrich®
L(+)-Histidine
Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 272…273 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
EINECS: 200-745-3
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM