L(-)-Tryptophan
Supplier: Molekula
Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
13831082-1KG 13831082-100G
MOLE13831082-1KGEA 412 EUR
MOLE13831082-1KG MOLE13831082-100G
L(-)-Tryptophan
L(-)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
Melting Pt: 290 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929