(R)-(-)-Hydratropic acid ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid
A17639.01
A17639.03
A17639.01EA
71.4
EUR
A17639.01
A17639.03
(R)-(-)-Hydratropic acid ≥97%
(R)-(-)-Hydratropic acid
Purity: 97%
CAS No.: 7782-26-5
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 148°(298°F)
Refractive Index: 1.5230
Melting Point: 32-34°
Boiling Point: 264-266°
Beilstein Reference: 2207688
Optical Rotation: -73° (c=1.6 in chloroform)
MDL No.: MFCD00063140
Formula:
C₉H₁₀O₂ Boiling Pt: 264…266 °C Melting Pt: 32…34 °C Density: 1.100 Flash Pt: 148 °C (298 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063140 CAS Number: 7782-26-5 |