D-(+)-Tyrosine, Sigma-Aldrich®
Supplier: Merck
Synonyms: H-D-Tyr-OH
M-Clarity™ quality level = MQ100
855456-500MG 855456-5G
SIAL855456-500MGEA 46.6 EUR
SIAL855456-500MG SIAL855456-5G
D-(+)-Tyrosine, Sigma-Aldrich®
D(+)-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 285
MDL Number: MFCD00063073
CAS Number: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970