L(-)-Tyrosine, Sigma-Aldrich®
Supplier: Merck
Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
M-Clarity™ quality level = MQ200
T3754-100G T3754-1KG T3754-500G T3754-50G
SIALT3754-100GEA 80.1 EUR
SIALT3754-100G SIALT3754-1KG SIALT3754-500G SIALT3754-50G
L(-)-Tyrosine, Sigma-Aldrich®
L(-)-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 285
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907