(1R,2R)-(-)-1,2-Diaminocyclohexane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (R,R)-DACH, (1R,2R)-Cyclohexane-1,2-diamine
M-Clarity™ quality level = MQ200
346721-1G 346721-5G
SIAL346721-1GEA 21.5 EUR
SIAL346721-1G SIAL346721-5G
(1R,2R)-(-)-1,2-Diaminocyclohexane, Sigma-Aldrich®
(1R,2R)-(-)-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 187
Melting Pt: 14…15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062985
CAS Number: 20439-47-8
UN: 3259
ADR: 8 II