(1S,2S)-(+)-trans-1,2-Cyclohexanediol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
421804-1G 421804-250MG
SIAL421804-1GEA 287 EUR
SIAL421804-1G SIAL421804-250MG
(1S,2S)-(+)-trans-1,2-Cyclohexanediol, Sigma-Aldrich®
(1S,2S)-(+)-trans-1,2-Cyclohexanediol
Formula: C₆H₁₂O₂
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 108 °C
CAS Number: 57794-08-8