D-(+)-Biotin, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
M-Clarity™ quality level = MQ200
14400-100MG
14400-1G
14400-5G
SIAL14400-100MGEA
32.9
EUR
SIAL14400-100MG
SIAL14400-1G
SIAL14400-5G
D-(+)-Biotin, Sigma-Aldrich®
D(+)-Biotin
Formula:
C₁₀H₁₆N₂O₃S Melting Pt: 232…233 °C Density: 1.41 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005541 CAS Number: 58-85-5 EINECS: 200-399-3 Merck Index: 13,01231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...