D-(+)-Biotin, Sigma-Aldrich®

Supplier: Merck

Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

M-Clarity™ quality level = MQ200

B4639-1G B4639-500MG B4639-5G
SIALB4639-1GEA 247 EUR
SIALB4639-1G SIALB4639-500MG SIALB4639-5G
D-(+)-Biotin, Sigma-Aldrich®
D(+)-Biotin
Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Melting Pt: 232…233 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR