5-Nitro-1,10-phenanthroline 98%
Supplier: Thermo Fisher Scientific
Warning
186080010 186080050
ACRO186080010EA 69.2 EUR
ACRO186080010 ACRO186080050
5-Nitro-1,10-phenanthroline 98%
5-Nitro-1,10-phenanthroline
Formula: C₁₂H₇N₃O₂
Melting Pt: 199…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00004981
CAS Number: 4199-88-6
EINECS: 224-097-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Light yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 199 to 203 °C
HPLC ≥97.5 %
Optical rotation -65° to -69° (1 % in MeOH)