1,8-Nonadiyne ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 97%
CAS No.: 2396-65-8
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 41°(105°F)
Refractive Index: 1.4490
Odour: Characteristic
Melting Point: -21°
Boiling Point: 55-56°/13mm
Beilstein Reference: 1733443
UN No.: 3295
MDL No.: MFCD00008581
Formula:
C₉H₁₂ Boiling Pt: 55…56 °C (13 mmHg) Melting Pt: –21 °C Density: 0.799 Flash Pt: 41 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008581 CAS Number: 2396-65-8 EINECS: 219-254-0 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...