1,2-Dibromoethane, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

D40752-1KG D40752-250G D40752-5G
SIALD40752-1KGEA 90.7 EUR
SIALD40752-1KG SIALD40752-250G SIALD40752-5G
1,2-Dibromoethane, Sigma-Aldrich®
1,2-Dibromoethane
Formula: BrH₂CCH₂Br
Boiling Pt: 131.7 °C (1013 hPa)
Melting Pt: 10 °C
Density: 2.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000233
CAS Number: 106-93-4
EINECS: 203-444-5
UN: 1605
ADR: 6.1,I
Merck Index: 13,03830

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR