1,3-Dibromopropane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
125903-1L 125903-50ML
SIAL125903-1LEA 587 EUR
SIAL125903-1L SIAL125903-50ML
1,3-Dibromopropane, Sigma-Aldrich®
1,3-Dibromopropane
Formula: BrH₂CCH₂CH₂Br
Boiling Pt: 167 °C (1013 hPa)
Melting Pt: –34 °C
Density: 1.99 g/cm³ (20 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00000255
CAS Number: 109-64-8
EINECS: 203-690-3
UN: 1993
ADR: 3,III
Merck Index: 12,09844