1-Bromopentadecane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
238333-10G 238333-50G
SIAL238333-10GEA 95.3 EUR
SIAL238333-10G SIAL238333-50G
1-Bromopentadecane, Sigma-Aldrich®
1-Bromopentadecane
Formula: BrH₂C(CH₂)₁₃CH₃
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 19 °C
Density: 1.0052 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000229
CAS Number: 629-72-1
EINECS: 211-104-2