1-Bromopropane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Solvent 38, n-Propyl bromide
M-Clarity™ quality level = MQ200
B78106-100ML B78106-1L B78106-500ML B78106-5ML
SIALB78106-100MLEA 34.8 EUR
SIALB78106-100ML SIALB78106-1L SIALB78106-500ML SIALB78106-5ML
1-Bromopropane, Sigma-Aldrich®
1-Bromopropane
Formula: BrH₂CCH₂CH₃
MW: 122.99 g/mol
Boiling Pt: 71
Melting Pt: –108 °C
Density: 1.354 g/cm³ (20 °C)
Flash Pt: 25
Storage Temperature: Ambient
MDL Number: MFCD00000254
CAS Number: 106-94-5
EINECS: 203-445-0
UN: 2344
ADR: 3 II
Merck Index: 12,08030
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63e928c183f1c573c1a2eba4ccb7f1147af103ed&vid=6b2b626154a7592a42b400fdf8ba1c1028a60850&prid=web&clid=SAPDAM