Bromoethane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Bromic ether, Hydrobromic ether
M-Clarity™ quality level = MQ200
124052-1L 124052-250ML
SIAL124052-1LEA 96.3 EUR
SIAL124052-1L SIAL124052-250ML
Bromoethane, Sigma-Aldrich®
Bromoethane
Formula: BrH₂CCH₃
MW: 108.97 g/mol
Boiling Pt: 38.4
Melting Pt: –118 °C
Density: 1.46 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00000232
CAS Number: 74-96-4
EINECS: 200-825-8
UN: 1891
ADR: 6.1 II
Merck Index: 13,03807