1,2-Dibromotetrachloroethane, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
133396-100G 133396-25G
SIAL133396-100GEA 113 EUR
SIAL133396-100G SIAL133396-25G
1,2-Dibromotetrachloroethane, Sigma-Aldrich®
1,2-Dibromotetrachloroethane
Formula: BrCl₂CCBrCl₂
MW: 325.64 g/mol
Boiling Pt: 222
Melting Pt: 83
Density: 2.713 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000780
CAS Number: 630-25-1
EINECS: 211-136-7