1-Bromo-4-chlorobutane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

B60800-100G B60800-25G
SIALB60800-100GEA 154 EUR
SIALB60800-100G SIALB60800-25G
1-Bromo-4-chlorobutane, Sigma-Aldrich®
1-Bromo-4-chlorobutane
Formula: BrH₂C(CH₂)₂CH₂Cl
Boiling Pt: 80 °C (1013 hPa)
Density: 1.4895 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001010
CAS Number: 6940-78-9
EINECS: 230-089-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR